Learn More
DuPont™ Tyvek™ 400 Shirt
GREENER_CHOICE
Catalog No. 17500281
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17500281 2X-Large
17500282 3X-Large
17500283 4X-Large
17500287 Large
17500288 Medium
17500289 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17500281 Supplier DuPont™ Supplier No. TY303SWH2X0012G1
400 shirts provide the protection and durability you need so you can focus on the task at hand. Meets USDA, FDA, OSHA Standards.
  • Shirt design with front snap closures for easy donning and doffing
  • Laydown collar
  • Open wrists
  • Typically worn with TY350S WH pants.
  • Manufactured under specifications that do not contain natural rubber latex
Compliance
USDA, FDA, OSHA
Specifications
Size 2X-Large
Gender Unisex
Material Tyvek
Color White
Neck Style Collar
Seam Type Serged
Closure Type Snaps
Cuff Style Open
Style Extends to Hip