Learn More
DuPont™ Tyvek™ 400 Shirt
GREENER_CHOICE
Catalog No. 17500284
Change view
Click to view available options
Size:
5X-Large
6X-Large
7X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17500284 5X-Large
17500285 6X-Large
17500286 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17500284 Supplier DuPont™ Supplier No. TY303SWH5X005000
400 shirts provide the protection and durability you need so you can focus on the task at hand.
  • Shirt design with front snap closures for easy donning and doffing
  • Laydown collar
  • Open wrists
  • Typically worn with TY350S WH pants
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Size 5X-Large
Gender Unisex
Material Tyvek
Color White
Neck Style Collar
Seam Type Serged
Closure Type Snaps
Cuff Style Open
Style Extends to Hip