Learn More
DuPont™ Tyvek™ 400 Pants
400 pants provide the protection and durability you need so you can focus on the task at hand. Meets USDA, FDA, OSHA Standards.
Supplier:  DuPont™ TY350SWHMD0012G1
 View more versions of this product
Catalog No. 17-500-297
Only null left
Explore available promotions
Description
Description
  • Pants with open ankle and elastic at waist for easy donning and removal
  • Suitable for applications that do not require the full body coverage of a coverall
  • Typically worn with TY303S WH shirt
  • Manufactured under specifications that do not contain natural rubber latex
  • Can be used in a variety of applications including general maintenance/operations, asbestos and lead abatement, mold remediation, and environmental cleanups
Compliance
USDA, FDA, OSHA
Specifications
Specifications
Medium
Tyvek
Open
Elastic
Unisex
White
Serged
Product Suggestions
Product Suggestions