missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 400 Frocks, Open Wrists
GREENER_CHOICE

Catalog No. 19820542
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
Packaging Type:
Bulk Packaged
Individually Packaged for Vending
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Packaging Type
19820542 X-Large Individually Packaged for Vending
19820541 Medium Individually Packaged for Vending
19820540 Large Individually Packaged for Vending
19820537 2X-Large Individually Packaged for Vending
19820538 3X-Large Individually Packaged for Vending
19820539 4X-Large Individually Packaged for Vending
17500259 5X-Large Bulk Packaged
17500261 7X-Large Bulk Packaged
17500260 6X-Large Bulk Packaged
19813470 4X-Large Bulk Packaged
19087225 3X-Large Bulk Packaged
19087224 2X-Large Bulk Packaged
0136111A Small Bulk Packaged
0136111B Medium Bulk Packaged
19087223 X-Large Bulk Packaged
0136111C Large Bulk Packaged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 19820542 Supplier DuPont™ Supplier No. TY210SWHXL0030VP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tyvek frock features a body-length jacket design with front snap closures for easy donning and doffing. Available in individually packaged styles for PPE vending machines.

  • Flash spun high density polyethylene nonwoven material
  • Provides ideal balance of protection, durability, and comfort
  • Offers an inherent barrier against particles (down to 1.0 micron in size)
  • Common applications include lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up
  • Frocks provide body-length jacket design with front snap closures for easy donning and doffing
  • Frock extends below hip
  • Laydown collar

Specifications

Size X-Large
Material Tyvek
Color White
Disposable Limited-use
Closure Type Snaps
Cuff Style Open
Collar Type Laydown
Style Below Hip-length
Size (English) Chest 41.75 to 45.25 in.
Certifications/Compliance ASTM D1777
Packaging Type Individually Packaged for Vending
Length (Metric) 99.06 cm
Length (English) 39 in.
Product Type General Purpose Frock
Seam Type Serged
For Use With (Application) Lead and asbestos abatement and remediation, general maintenance and operations, spray painting, general clean-up
Show More Show Less