missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 400 Coveralls with Elastic Wrists and Ankles, Vending Packaged
GREENER_CHOICE

Catalog No. p-200003807
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820526 Medium
19820525 Large
19820527 X-Large
19820519 2X-Large
19820520 3X-Large
19820521 4X-Large
19820522 5X-Large
19820523 6X-Large
19820524 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19820526 Supplier DuPont™ Supplier No. TY125SWHMD0025VP
Only null left

Individually packaged coveralls for PPE vending feature elastic wrists and ankles to provide an ideal balance of protection, durability, and comfort.

  • Vending packaged, serged seams
  • Tyvek composed of flash spun high density polyethylene which creates a unique, nonwoven material that provides durability and comfort
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Laydown collar; longer zipper extends to chin for complete coverage of neck area
  • Storm flap of garment material covers zipper
  • Elastic opening for tighter fit at wrists and ankles
  • Elastic waist to better position the garment
  • Thumb loop to keep garment sleeve in place
  • Applications include fiberglass and composite manufacturing, wind turbines, boat construction and repair, remediation, and maintenance

Specifications

Product Type General Purpose Coveralls
Size Medium
Gender Unisex
Material Tyvek
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle
Collar Type Laydown
Certifications/Compliance ASTM D1777, D3776, D774, D5733, D5034, D257, AATCC 127
Description Flashspun HDPE
Latex Free Yes
Model TY125SWH
For Use With (Application) Lead and Asbestos Abatement, Remediation, General Maintenance, Operations, Spray Painting, Cleaning
Quantity 25/Cs.
Packaging Individually Packaged for PPE Vending
Show More Show Less