missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 400 Coveralls with Respirator Fit Hood, Elastic Wrists, USDA, FDA, OSHA
GREENER_CHOICE

Catalog No. 17500197
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17500197 2X-Large
17500199 3X-Large
17500201 4X-Large
17500206 Large
17500208 Medium
17500210 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17500197 Supplier DuPont™ Supplier No. TY122SWH2X0006G1

Coveralls with attached skid-resistant boots include stormflaps and a respirator fit hood. Meets USDA, FDA, OSHA Standards.

  • Comfort fit design based on extensive wearer input, meets USDA, FDA, and OSHA standards
  • Enables a greater range of movement while stretching and bending
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Longer zipper extends to chin for complete coverage of neck area
  • Elastic opening for tighter fit at wrist
  • Attached Tyvek™ boots have special coating to provide added skid-resistance
  • 25 Coveralls/Cs.

Compliance

USDA, FDA, OSHA

Specifications

Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material Tyvek
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic, Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic, Respirator Fit
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle
Model TY122SWH
For Use With (Application) Lead and Asbestos Abatement, Remediation, General Maintenance and Operations, Spray Painting, General Cleaning
Quantity 25/Cs.
Show More Show Less