missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 400 Hooded Coveralls with Skid-Resistant Boots, USMCA Sourced
GREENER_CHOICE

Catalog No. 17500200
Change view
Click to view available options
Size:
3X-Large
5X-Large
6X-Large
7X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17500200 3X-Large
17500203 5X-Large
17500204 6X-Large
17500205 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17500200 Supplier DuPont™ Supplier No. TY122SWH3X0025NF

Coveralls with attached skid-resistant boots include stormflaps and a respirator fit hood. Meets NAFTA Standards.

  • Comfort fit design based on extensive wearer input, meets NAFTA standards
  • Enables a greater range of movement while stretching and bending
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Longer zipper extends to chin for complete coverage of neck area
  • Elastic opening for tighter fit at wrist
  • Attached Tyvek™ boots have special coating to provide added skid-resistance
  • 25/Cs.

Compliance

NAFTA

Specifications

Product Type General Purpose Coveralls
Size 3X-Large
Gender Unisex
Material Tyvek
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic, Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic, Respirator Fit
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle
Certifications/Compliance Meets NAFTA Standards
Model TY122SWH
For Use With (Application) Lead and Asbestos Abatement, Remediation, General Maintenance and Operations, Spray Painting, General Cleaning
Quantity 25/Cs.
Show More Show Less