missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 400 Hooded Coverall
GREENER_CHOICE

Catalog No. 17500233
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17500233 2X-Large
17500235 3X-Large
17500237 4X-Large
17500243 Large
17500245 Medium
17500247 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17500233 Supplier DuPont™ Supplier No. TY127SWH2X0006G1

Coveralls with respirator fit hood provide breathable inherent barrier protection against hazardous dry particles, aerosols, and non-hazardous light liquid splash. Meets NAFTA Standards. Meets USDA, FDA, OSHA Standards.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Comfort fit design based on extensive wearer input to provide our most comfortable garment design that: enables a greater range of movement while stretching and bending, provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts, utilizes a longer zipper for easier donning and doffing and an elastic waist to better position the garment
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Longer zipper extends to chin for complete coverage of neck area
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Stormflaps.

Specifications

Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material Tyvek
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Respirator Fit
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Certifications/Compliance ASTM D1777, D3776, D774, D5733, D5034, D257, AATCC 127
Latex Free Yes
Model TY127SWH
For Use With (Application) Lead and Asbestos Abatement, Remediation, General Maintenance, Operations, Spray Painting, Cleaning
Height (English) 6 to 6.4 ft.
Quantity 6/Cs.
Show More Show Less