Learn More
DuPont™ Tyvek™ 400 Coveralls, 125S Series, Elastic Wrists and Ankles, NAFTA
Coveralls with elastic wrists and ankles provide an ideal balance of protection, durability, and comfort.
Supplier:  DuPont™ TY125SWHMD0025NF
 View more versions of this product
Catalog No. 17-500-230
Only null left
Explore available promotions
Description
Description
  • USMCA sourced, serged seams
  • Tyvek composed of flash spun high density polyethylene which creates a unique, nonwoven material that provides durability and comfort
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Laydown collar; longer zipper extends to chin for complete coverage of neck area
  • Storm flap of garment material covers zipper
  • Elastic opening for tighter fit at wrists and ankles
  • Elastic waist to better position the garment
  • Thumb loop to keep garment sleeve in place
  • Applications include fiberglass and composite manufacturing, wind turbines, boat construction and repair, remediation, and maintenance
Compliance
NAFTA
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Elastic
Unhooded
Serged
Particle
ASTM D1777, D3776, D774, D5733, D5034, D257, AATCC 127
Yes
Lead and Asbestos Abatement, Remediation, General Maintenance, Operations, Spray Painting, Cleaning
Medium
Tyvek
Limited-use
Elastic
Collared
Front Zipper
Non-sterile
Laydown
Flashspun HDPE
TY125SWH
25/Cs.
Product Suggestions
Product Suggestions