missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 400 Coveralls, 125S Series, Elastic Wrists and Ankles, NAFTA
GREENER_CHOICE

Catalog No. 17500226
Change view
Click to view available options
Size:
Medium
Large
X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17500226 7X-Large
17500223 4X-Large
17500228 Large
17500221 3X-Large
17500232 X-Large
17500225 6X-Large
17500230 Medium
17500224 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17500226 Supplier DuPont™ Supplier No. TY125SWH7X0025NF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Coveralls with elastic wrists and ankles provide an ideal balance of protection, durability, and comfort.

  • USMCA sourced, serged seams
  • Tyvek composed of flash spun high density polyethylene which creates a unique, nonwoven material that provides durability and comfort
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Laydown collar; longer zipper extends to chin for complete coverage of neck area
  • Storm flap of garment material covers zipper
  • Elastic opening for tighter fit at wrists and ankles
  • Elastic waist to better position the garment
  • Thumb loop to keep garment sleeve in place
  • Applications include fiberglass and composite manufacturing, wind turbines, boat construction and repair, remediation, and maintenance

Compliance

NAFTA

Specifications

Product Type General Purpose Coveralls
Size 7X-Large
Gender Unisex
Material Tyvek
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle
Collar Type Laydown
Certifications/Compliance ASTM D1777, D3776, D774, D5733, D5034, D257, AATCC 127
Description Flashspun HDPE
Latex Free Yes
Model TY125SWH
For Use With (Application) Lead and Asbestos Abatement, Remediation, General Maintenance, Operations, Spray Painting, Cleaning
Quantity 25/Cs.
Show More Show Less