missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 400 Hooded Coverall

Catalog No. 19812796
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19812796 4X-Large
17500241 7X-Large
013617B Medium
013617C Large
013617D X-Large
013617E 2X-Large
013617F 3X-Large
19120593 5X-Large
191303906 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19812796 Supplier DuPont™ Supplier No. TY127SWH4X002500

Coveralls with respirator fit hood provide breathable inherent barrier protection against hazardous dry particles, aerosols, and non-hazardous light liquid splash.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Comfort fit design based on extensive wearer input to provide our most comfortable garment design that: enables a greater range of movement while stretching and bending, provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts, utilizes a longer zipper for easier donning and doffing and an elastic waist to better position the garment
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Longer zipper extends to chin for complete coverage of neck area
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Stormflaps.

Compliance

Meets USDA, FDA, OSHA standards; ASTM™ D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127 compliant.

Specifications

Product Type General Purpose Coveralls
Size 4X-Large
Gender Unisex
Material Tyvek
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Respirator Fit
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Certifications/Compliance USDA, FDA, OSHA, ASTM D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127
Latex Free Yes
Model TY127SWH
Size (English) Chest 52.75 to 56.25 in.
Size (English) Inseam 33 in.
Size (Metric) Inseam 83.82 cm
For Use With (Application) Lead and Asbestos Abatement, Remediation, General Maintenance, Operations, Spray Painting, Cleaning
Length (English) Sleeve 39 in.
Length (Metric) Sleeve 99.06 cm
Quantity 25/Cs.
Show More Show Less