Learn More
DuPont™ Tyvek™ 400 Hooded Coverall
Coveralls with respirator fit hood provide breathable inherent barrier protection against hazardous dry particles, aerosols, and non-hazardous light liquid splash.
Supplier:  DuPont™ TY127SWH5X002500
 View more versions of this product
Catalog No. 19-120-593
Only null left
Explore available promotions
Description
Description
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Comfort fit design based on extensive wearer input to provide our most comfortable garment design that: enables a greater range of movement while stretching and bending, provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts, utilizes a longer zipper for easier donning and doffing and an elastic waist to better position the garment
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Longer zipper extends to chin for complete coverage of neck area
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Stormflaps.
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Elastic
Respirator Fit
Serged
ASTM D1777, D3776, D774, D5733, D5034, D257, AATCC 127
TY127SWH
34 in.
86.36 cm
Lead and Asbestos Abatement, Remediation, General Maintenance, Operations, Spray Painting, Cleaning
101.6 cm
2.04 to 1.85 m
5X-Large
Tyvek
Limited-use
Elastic
Hooded
Front Zipper
Non-sterile
Yes
55.75 to 59.25 in.
141.6 to 150.5 cm
5.7 mil
40 in.
6.7 to 6.10 ft.
25/Cs.
Product Suggestions
Product Suggestions