missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin B46, 98+%, Thermo Scientific Chemicals

Catalog No. AAJ61704MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61704MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61704MA Supplier Thermo Scientific Chemicals Supplier No. J61704MA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibitor of EGF receptor kinase activity

Inhibitor of EGF receptor kinase activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 133550-34-2
Molecular Formula C19H18N2O3
Molecular Weight (g/mol) 322.364
MDL Number MFCD00209865
InChI Key GSQOBTOAOGXIFL-WJDWOHSUSA-N
Synonym tyrphostin b46, z-2-cyano-3-3,4-dihydroxyphenyl-n-3-phenylpropyl prop-2-enamide, n-3 inverted exclamation marka-phenylpropyl-3,4-dihydroxybenzylidenecyanoacetamide
PubChem CID 5353390
IUPAC Name (Z)-2-cyano-3-(3,4-dihydroxyphenyl)-N-(3-phenylpropyl)prop-2-enamide
SMILES C1=CC=C(C=C1)CCCNC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N

Specifications

Color Yellow
Quantity 10 mg
Formula Weight 322.36
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Tyrphostin B46
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only