missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin AG 494 98.0+%, TCI America™

Catalog No. p-7118031
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A270420MG 20 mg
A2704100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A270420MG Supplier TCI America Supplier No. A270420MG
Only null left

Chemical Identifiers

CAS 133550-35-3
Molecular Formula C16H12N2O3
Molecular Weight (g/mol) 280.283
MDL Number MFCD00209863
InChI Key HKHOVJYOELRGMV-XYOKQWHBSA-N
Synonym tyrphostin b48, tyrphostin ag 494, tyrphostin ag-494, e-2-cyano-3-3,4-dihydroxyphenyl-n-phenylacrylamide, alpha-cyano-3,4-dihydroxy-n-phenylcinnamide, chembl56731, 2e-2-cyano-3-3,4-dihydroxyphenyl-n-phenylprop-2-enamide, e-2-cyano-3-3,4-dihydroxyphenyl-n-phenyl-2-propenamide, n-phenyl-3,4-dihydroxybenzylidenecyanoacetamide, tocris-0619
PubChem CID 5328771
IUPAC Name (E)-2-cyano-3-(3,4-dihydroxyphenyl)-N-phenylprop-2-enamide
SMILES C1=CC=C(C=C1)NC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N

Specifications

Melting Point 249°C
Quantity 20 mg
UN Number 3439
Formula Weight 280.28
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Tyrphostin AG 494
TSCA No
Recommended Storage Refrigerator