missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin AG 490 98.0+%, TCI America™

Catalog No. T296220MG
Change view
Click to view available options
Quantity:
20 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T296220MG 20 mg
T2962100MG 100 mg
T29621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T296220MG Supplier TCI America Supplier No. T296220MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 133550-30-8
Molecular Formula C17H14N2O3
Molecular Weight (g/mol) 294.31
InChI Key TUCIOBMMDDOEMM-RIYZIHGNSA-N
PubChem CID 5328779
IUPAC Name (E)-N-benzyl-2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enamide
SMILES C1=CC=C(C=C1)CNC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N

Specifications

Quantity 20 mg
Synonym tyrphostin b42, tyrphostin ag 490, e-n-benzyl-2-cyano-3-3,4-dihydroxyphenyl acrylamide, tyrphostin ag490, tyrphostin ag-490, alpha-cyano-3,4-dihydroxy-n-benzylcinnamide, 2e-n-benzyl-2-cyano-3-3,4-dihydroxyphenyl prop-2-enamide, ag-490 tyrphostin b42, n-benzyl-2-cyano-3-3,4-dihydroxyphenyl acrylamide, chembl56543