missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tyrode's Solution, HEPES-buffered

Catalog No. AAJ67607AP
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67607AP 500 mL
AAJ67607K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67607AP Supplier Thermo Scientific Chemicals Supplier No. J67607AP

A solution for cell washing

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Tyrode's Solution
Packaging Plastic bottle
Quantity 500 mL
Physical Form Liquid

RUO – Research Use Only