missing translation for 'onlineSavingsMsg'
Learn More

Tyramine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7116819
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A03031G 1 g
A030325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A03031G Supplier TCI America Supplier No. A03031G
Only null left

Chemical Identifiers

CAS 60-19-5
Molecular Formula C8H12ClNO
Molecular Weight (g/mol) 173.64
MDL Number MFCD00012901
InChI Key RNISDHSYKZAWOK-UHFFFAOYSA-N
Synonym tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147
PubChem CID 66449
IUPAC Name hydrogen 4-(2-aminoethyl)phenol chloride
SMILES [H+].[Cl-].NCCC1=CC=C(O)C=C1

Specifications

Melting Point 269°C
Color White-Yellow
Quantity 1 g
Formula Weight 173.64
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tyramine Hydrochloride
RTECSNumber SJ6050000
TSCA Yes