missing translation for 'onlineSavingsMsg'
Learn More

Tyramine, 97%

Catalog No. p-210251
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC140610050 5 g Glass bottle
AC140610250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC140610050 Supplier Thermo Scientific Chemicals Supplier No. 140610050
Only null left

CAS: 51-67-2 | C8H11NO | 137.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51-67-2
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00008193
InChI Key DZGWFCGJZKJUFP-UHFFFAOYSA-N
Synonym tyramine, 4-2-aminoethyl phenol, 4-hydroxyphenethylamine, p-tyramine, 2-4-hydroxyphenyl ethylamine, uteramine, tyramin, tyrosamine, tocosine, 4-hydroxyphenylethylamine
PubChem CID 5610
ChEBI CHEBI:15760
IUPAC Name 4-(2-aminoethyl)phenol
SMILES C1=CC(=CC=C1CCN)O

Specifications

Melting Point 158.0°C to 163.0°C
Color Brown to Yellow
Boiling Point 175.0°C to 181.0°C (8.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula HOC6H4CH2CH2NH2
Quantity 5 g
Beilstein 13, 625
Merck Index 15, 10016
Solubility Information Solubility in water: 1g/95mL (15°C). Other solubilities: 1g/10mL boiling alcohol, sparingly soluble in benzene and xylene, soluble in ethanol
Formula Weight 137.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Tyramine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-115-8
RTECSNumber SJ5950000

RUO – Research Use Only