Learn More
Thermo Scientific™ Deionized Water
Catalog No. p-4528042
Change view
Click to view available options
Quantity:
1 gal.
5 gal.
12 x 16 oz.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23751610 1 gal.
23290065 12 x 16 oz.
23751628 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23751610 Supplier Thermo Scientific™ Supplier No. 751 610
Only null left
Thermo Scientific™ Deionized Water.
Ozonated to inhibit microbial growth.
  • For use in flame photometry, blood gas humidifying chambers, electrode maintenance and instrument cleaning
Note:
Because of the many ways customers use this product, Thermo Fisher Scientific cannot provide an open container stability claim. Customers may establish their own open container stability claim by following CLSI guidelines.
Specifications
Quantity 1 gal.
Not intended for infusion, injection or irrigation.