missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyloxapol, USP, Spectrum™ Chemical
Description
Spectrum™ Chemical Tyloxapol, USP
Specifications
Specifications
| CAS Max % | 1 |
| pH | 4.0 to 7.0 |
| Ignition Residue | 0.01 |
| Packaging | Amber Glass Bottle |
| Quantity | 25 g |
| Grade | USP |