missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 4000 Hooded Coverall

Catalog No. p-6926035
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Seam Type:
Taped
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Seam Type
19088348B Medium Taped
19088348C Large Taped
19088348D X-Large Taped
19088348E 2X-Large Taped
19088348F 3X-Large Taped
19088348G 4X-Large Taped
19162771 5X-Large Taped
19162773 6X-Large Taped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 19088348B Supplier DuPont™ Supplier No. SL122TWHMD000600
Only null left
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Delivers effective protection against a range of chemical environments.

Tychem SL, utilizes Saranex™ 23-P film laminated to Tyvek fabric to provide a rugged and durable fabric that resists abrasion. The fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.

  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Attached hood with elastic around face opening
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material

Compliance

Meets ASTM™ D3776, ASTM D1777, ASTM D3786, ASTM D5034 standards.

Specifications

Product Type Hooded Coveralls with Socks
Size Medium
Gender Unisex
Material Tychem
Color White
Disposable Single-use
Ankle Style Attached Socks
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type Front Zipper
Seam Type Taped
Sterility Non-sterile
Description Attached hood, elastic wrist, attached socks
Size (English) Chest 35.25 to 38 .75 in.
Size (English) Inseam 27.5 in.
Size (Metric) Inseam 69.85 cm
Thickness 12 mil
For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
Length (English) Sleeve 33.75 in.
Length (Metric) Sleeve 85.72 cm
Width (English) Chest 24.25 in.
Width (Metric) Chest 61.59 cm
Height (English) 5.25 to 5.58 ft.
Height (Metric) 1.58 to 1.7 m
Show More Show Less