Learn More
DuPont™ Tychem™ RESPONDER™ CSM Encapsulated Level A Suit with MSA CAMDS Connector
CSM Encapsulated Level A Suit is a front entry, low visibility suit with attached socks and gloves
Supplier:  DuPont™ RC550TTNMD00017C
 View more versions of this product
Catalog No. 18-888-744
Only null left
Explore available promotions
Description
Description
  • CSM Encapsulated Level A Suit with MSA CAMDS Connector design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment
  • Two exhaust valves release higher pressure inside encapsulating garments while simultaneously helping to prevent inward leakage of external vapors or particles into the garment. One-way valves are positive pressure and open on demand. Valves include splash covers made of barrier material to help prevent liquid intrusion. Valves are located on left back of head and right lower back
  • Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • Double taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered, both inside and outside of garment with compatible chemical-resistant material by heat-sealing.
  • Standard-width, anti-fog, three-layer (PVC 40 mil / Teflon™5 mil / PVC 20 mil) faceshield
  • Front entry design allows wearer to partially don garment (maintain standby position) without assistance
  • Expanded back to accommodate self-contained air breathing apparatus (SCBA)
  • Vapor tight zipper. Extra long to increase garment opening and aid donning and doffing
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
Specifications
Specifications
Tan
Medium
Zipper
Tychem
Boot Overflaps
Taped
Product Suggestions
Product Suggestions