Learn More
DuPont™ Tychem™ RESPONDER™ CSM Encapsulated Level A Suit with Interspiro
CSM Encapsulated Level A Suit is a front entry, low visibility suit with attached socks and gloves
Supplier:  DuPont™ RC550TTNMD00017W
 View more versions of this product
Catalog No. 18888746
Only null left
Explore available promotions
Description
Description
  • CSM Encapsulated Level A Suit design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment
  • Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • Double taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered, both inside and outside of garment with compatible chemical-resistant material by heat-sealing
  • Standard-width, anti-fog, three-layer (PVC 40 mil / Teflon™ 5 mil / PVC 20 mil) faceshield
  • Front entry design allows wearer to partially don garment (maintain standby position) without assistance
  • Expanded back to accommodate self-contained air breathing apparatus (SCBA)
  • Vapor tight zipper. Extra long to increase garment opening and aid donning and doffing
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
Specifications
Specifications
Tan
Medium
Zipper
Tychem
Boot Overflaps
Taped
Product Suggestions
Product Suggestions