missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 6000 SFR FS275T LT Apron
Description
- Tychem™ 6000 SFR garments provide broad chemical protection and secondary flame resistance in a durable, lightweight garment for use in environments where chemical splash and flash fire hazards exist.
- Designed to be worn over primary flame-resistant protective clothing, Tychem™ 6000 SFR garments help provide industrial workers and hazmat responders with permeation protection against a range of chemicals while providing escape time from flash fires.
- Tychem™ 6000 SFR fabric provides at least 30 min. of protection for more than 250 chemical challenges including chemical warfare agents, toxic industrial chemicals, and flammable organic solvents.
- In the event that a flash fire occurs, Tychem™ 6000 SFR garments won't contribute to additional burn injury if appropriate primary flame-resistant personal protective equipment is worn beneath.
Specifications
Specifications
| Type | Apron |
| Size | Small |
| Material | Tychem |
| Color | Tan |
| Disposable | Single-use |
| Closure Type | Hook and Loop, Waist Ties |
| Sterility | Non-sterile |
| Latex Free | No |
| No. of Pockets | None |
| Seam Type | Taped |