Learn More
DuPont™ Tychem™ 6000 Coverall with Respirator Fit Hood
6000 Coverall with respirator fit hood provide protection from chemical warfare agents and toxic industrial chemicals.
Supplier:  DuPont™ TF199TGYMD0006WG
 View more versions of this product
Catalog No. 18-888-799
Only null left
Explore available promotions
Description
Description
  • Certified to NFPA 1990 (NFPA 1992), 2022 edition
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Hooded coverall with longer zipper that extends to chin for complete coverage of neck area and good fit around a respirator
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Attached Dual Layer Gloves, Internal: Multi-layer laminate / External: Butyl
  • Attached socks
  • Outer boot flaps
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Specifications
Gray
Medium
Zipper
Single-use
Yes
None
Liquid
Non-sterile
Tychem
Boot Overflaps
Attached Gloves
Unisex
Hooded
6 Pk.
Taped
Product Suggestions
Product Suggestions