Learn More
DuPont™ Tychem™ 5000 Jacket
Catalog No. p-7139030
Change view
Click to view available options
Size:
Small
2X-Large
3X-Large
4X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17500122 2X-Large
17500124 3X-Large
17500126 4X-Large
17500130 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17500122 Supplier DuPont™ Supplier No. C3670TTN2X000600
Only null left
DuPont™ Tychem™ 5000 Jacket with Mandarin Collar, Elastic Wrists provides protection against a broad range of chemicals and situations involving liquid splash.
  • Ideal for applications that do not require the full body coverage of a coverall
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
  • Mandarin collar designed to fit tightly around hooded PAPR
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Typically worn with C3360T TN pants or sold as jacket/pant ensemble (C3750T TN)
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks
Specifications
Size 2X-Large
Gender Unisex
Material Tychem
Color Tan
Closure Type Zipper
Collar Type Mandarin Collar
Cuff Style Elastic
Seam Type Taped