missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 5000 Encapsulated Level B Suit

Catalog No. 17500119
Change view
Click to view available options
Size:
Small
Medium
Large
4X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17500119 Small
17500116 4X-Large
17500117 Large
17500118 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17500119 Supplier DuPont™ Supplier No. C3526TTNSM000600

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rear entry provides protection against a broad range of chemicals and situations involving liquid splash

  • Encapsulated design provides protection from liquid splash exposure for both the wearer and respiratory equipment
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks
  • Also provides coverage from overhead liquid splash (does not provide vapor protection)
  • Taped seams provide strong chemical resistance against heavy liquid splashes
    • Sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
  • Standard 20 mil PVC face shield
  • Rear entry design keeps closure away from direct contact with frontal chemical exposure
  • Flat back design for use with airline
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material

Specifications

Color Tan
For Use With (Application) Chemical Handling, Hazmat, Emergency Response, Industrial Applications
Material Tychem
Size Small
Ankle Style Boot Overflaps
Closure Type Zipper
Product Type Encapsulated Level B Suit
Seam Type Taped