Learn More
DuPont™ Tychem™ 5000 Encapsulated Level B
Level B Suit, rear entry provides protection against a broad range of chemicals and situations involving liquid splash.
Supplier:  DuPont™ C3526TTNLG000600
 View more versions of this product
Catalog No. 17-500-117
Only null left
Explore available promotions
Description
Description
  • Encapsulated design provides protection from liquid splash exposure for both the wearer and respiratory equipment
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
  • Also provides coverage from overhead liquid splash (does not provide vapor protection)
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Standard 20 mil PVC faceshield
  • Rear entry design keeps closure away from direct contact with frontal chemical exposure
  • Flat back design for use with airline
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
Specifications
Specifications
Tan
Large
Zipper
Tychem
Boot Overflaps
Taped
Product Suggestions
Product Suggestions