missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 5000 Encapsulated Level B Suit

Catalog No. 17500115
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17500115 X-Large
17500109 2X-Large
17500110 3X-Large
17500111 4X-Large
17500112 Large
17500113 Medium
17500114 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17500115 Supplier DuPont™ Supplier No. C3525TTNXL000600

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Front entry provides protection against a broad range of chemicals and situations involving liquid splash

  • Encapsulated design provides protection from liquid splash exposure for both the wearer and respiratory equipment
  • Also provides coverage from overhead liquid splash (does not provide vapor protection)
  • Taped seams provide strong chemical resistance against heavy liquid splashes
  • Standard 20 mil PVC face shield
  • Front entry design allows wearer to partially don garment (maintain standby position) without assistance
  • Flat back design for use with airline
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Manufactured under specifications that do not contain natural rubber latex

Specifications

Color Tan
For Use With (Application) Chemical Handling, Hazmat, Law Enforcement, Domestic Preparedness
Material Tychem
Size X-Large
Ankle Style Boot Overflaps
Closure Type Zipper
Product Type Encapsulated Level B Suit
Seam Type Taped