Learn More
DuPont™ Tychem™ 5000 Encapsulated Level B
Encapsulated Level B Suit, Front Entry provides protection against a broad range of chemicals and situations involving liquid splash.
Supplier: DuPont™ C3525TTNMD000600
Description
- Encapsulated design provides protection from liquid splash exposure for both the wearer and respiratory equipment
- Also provides coverage from overhead liquid splash (does not provide vapor protection)
- Taped seams provide strong chemical resistance against heavy liquid splashes
- Standard 20 mil PVC faceshield
- Front entry design allows wearer to partially don garment (maintain standby position) without assistance
- Flat back design for use with airline
- Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Manufactured under specifications that do not contain natural rubber latex
Specifications
| Tan | |
| Medium | |
| Zipper |
| Tychem | |
| Boot Overflaps | |
| Taped |