Learn More
DuPont™ Tychem™ 5000 Encapsulated Level B
Catalog No. 17500109
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17500109 2X-Large
17500110 3X-Large
17500111 4X-Large
17500112 Large
17500113 Medium
17500114 Small
17500115 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17500109 Supplier DuPont™ Supplier No. C3525TTN2X000600
Encapsulated Level B Suit, Front Entry provides protection against a broad range of chemicals and situations involving liquid splash.
  • Encapsulated design provides protection from liquid splash exposure for both the wearer and respiratory equipment
  • Also provides coverage from overhead liquid splash (does not provide vapor protection)
  • Taped seams provide strong chemical resistance against heavy liquid splashes
  • Standard 20 mil PVC faceshield
  • Front entry design allows wearer to partially don garment (maintain standby position) without assistance
  • Flat back design for use with airline
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Color Tan
Material Tychem
Size 2X-Large
Ankle Style Boot Overflaps
Closure Type Zipper
Seam Type Taped