Learn More
DuPont™ Tychem™ 5000 Coverall (all)
DuPont™ Tychem™ 5000 Coverall with Collar, Elastic Wrists and Ankles provides protection against a broad range of chemicals and situations involving liquid splash.
$505.99 - $505.99
Specifications
Gender Unisex
Material Tychem
Color Tan
Ankle Style Elastic
Cuff Style Elastic
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-500-038
View Documents
DuPont™
C3125TTNMD000600
Medium
Case of 6 Each for $505.99
Only null left
17-500-039
View Documents
DuPont™
C3125TTNSM000600
Small
Case of 6 Each for $505.99
Only null left
Description
Description
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Laydown collar
  • Strong zipper for tough, flexible closure
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Specifications
Specifications
Unisex
Tan
Elastic
Taped
Tychem
Elastic
Zipper
Collared
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'