Learn More
DuPont™ Tychem™ 5000 Bib Overall
Description
- Bib style overall with adjustable webbing straps
- Typically worn in combination with C3670T TN jacket or in applications were potential exposure is below waist
- Manufactured under specifications that do not contain natural rubber latex
- Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks
- NFPA 1992 certified
Specifications
Specifications
| Closure Type | Adjustable Webbing Straps with Snap-lock |
| Color | Tan |
| Material | Tychem |
| Seam Type | Taped |
| Size | Small |
| Suspender | Adjustable Webbing Straps with Buckle Closure |
| Ankle Style | Open |
| Product Type | Protective Bib Overall |