missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 5000 Bib Overall And Jacket Combo

Catalog No. 17500132
Change view
Click to view available options
Size:
4X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17500132 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17500132 Supplier DuPont™ Supplier No. C3750TTN4X000600

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides protection against a broad range of chemicals and situations involving liquid splash

  • Jacket and bib overall combination ensemble
    • Components also sold separately
  • Comes with a jam fit cuff and glove insert for user supplied gloves
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.

Specifications

Color Tan
Hood Type None
Material Tychem
Size 4X-Large
Ankle Style Open
Closure Type Zipper
Cuff Style Jam Fit
Product Type Bib Overall and Jacket Combo
Seam Type Taped
Suspender Adjustable Webbing Straps with Buckle Closure