Learn More
DuPont™ Tychem™ 4000 Hooded Coverall
Ideal for chemical mixing, remediation, emergency medical response, paint spraying, and radioactive environments
Supplier:  DuPont™ SL127TWHMD000600
 View more versions of this product
Catalog No. 19-088-2132
Only null left
Explore available promotions
Alert:
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Attached hood with elastic around face opening.
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Compliance
Meets ASTM™ D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786; 16 CFR 1610 Standard for Flammability of Clothing Textiles; MIL-STD-282 requirements.
Specifications
Specifications
Medium
Tychem
Elastic
Hooded
Taped
28 in.
12 mil
33.75 in.
24.25 in.
1.6 to 1.73 m
0.1 kg
Unisex
White
Elastic
Front Zipper
ASTM D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786, 16 CFR 1610, MIL-STD-282
89.53 to 98.42 cm
Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
85.72 cm
5.3 to 5.7 ft.
0.22 lb.
Product Suggestions
Product Suggestions