missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 4000 Hooded Coverall

Catalog No. 190882005B
Change view
Click to view available options
Closure Type:
Front Zipper
Seam Type:
Taped
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Height (English):
5.3 to 5.7 ft.
5.5 to 5.9 ft.
5.8 to 6.2 ft.
6 to 6.4 ft.
6.2 to 6.4 ft.
6.4 to 6.7 ft.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type Seam Type Size Height (English)
190882005B Front Zipper Taped 3X-Large 6.2 to 6.4 ft.
19087152 Front Zipper Taped 2X-Large 6 to 6.4 ft.
190882005A Front Zipper Taped X-Large 5.8 to 6.2 ft.
190882132 Front Zipper Taped Medium 5.3 to 5.7 ft.
190882131 Front Zipper Taped Large 5.5 to 5.9 ft.
190882134 Front Zipper Taped 4X-Large 6.4 to 6.7 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 190882005B Supplier DuPont™ Supplier No. SL127TWH3X000600
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Ideal for chemical mixing, remediation, emergency medical response, paint spraying, and radioactive environments

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Attached hood with elastic around face opening.
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.

Compliance

Meets ASTM™ D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786; 16 CFR 1610 Standard for Flammability of Clothing Textiles; MIL-STD-282 requirements.

Specifications

Size 3X-Large
Gender Unisex
Material Tychem
Color White
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Front Zipper
Seam Type Taped
Certifications/Compliance ASTM D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786, 16 CFR 1610, MIL-STD-282
Size (English) Inseam 31.5 in.
Size (Metric) Chest 120 to 128.9 cm
Thickness 12 mil
For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
Length (English) Sleeve 38.5 in.
Length (Metric) Sleeve 97.79 cm
Width (English) Chest 30.25 in.
Height (English) 6.2 to 6.4 ft.
Height (Metric) 1.88 to 1.95 m
Weight (English) 0.22 lb.
Weight (Metric) 0.1 kg
Show More Show Less