missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 4000 Hooded Coverall
Ideal for chemical mixing, remediation, emergency medical response, paint spraying, and radioactive environments
Supplier: DuPont™ SL127TWH2X000600
Alert:
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
- Attached hood with elastic around face opening.
- Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Elastic opening for tighter fit at ankle
- Manufactured under specifications that do not contain natural rubber latex
- Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
Compliance
Meets ASTM™ D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786; 16 CFR 1610 Standard for Flammability of Clothing Textiles; MIL-STD-282 requirements.
Specifications
| 2X-Large | |
| Tychem | |
| Elastic | |
| Hooded | |
| Taped | |
| 30.5 in. | |
| 12 mil | |
| 38.25 in. | |
| 28.75 in. | |
| 1.8 m | |
| 0.1 kg |
| Unisex | |
| White | |
| Elastic | |
| Front Zipper | |
| ASTM D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786, 16 CFR 1610, MIL-STD-282 | |
| 112.39 to 121.28 cm | |
| Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments | |
| 97.15 cm | |
| 6 to 6.4 ft. | |
| 0.22 lb. |