missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 4000 Coverall (all)
Useful when handling many wet and dry chemicals, and certain hazardous materials such as PCBs - See more at: http://beta.fishersci.com/shop/products//p-25013#sthash.pbR5bBoS.dpuf
Supplier: DuPont™ SL125BWH2X001200
Description
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
- Laydown collar
- Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Elastic opening for tighter fit at ankle
- Manufactured under specifications that do not contain natural rubber latex
Compliance
Meets ASTM™ D3776, ASTM D1777, ASTM D3786, ASTM D5034 standards.
Specifications
| 2X-Large | |
| Tychem | |
| Elastic | |
| Collared | |
| Bound | |
| 30 in. | |
| Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments | |
| 97.15 cm | |
| 6 to 6.4 ft. |
| Unisex | |
| White | |
| Elastic | |
| Front Zipper | |
| Elastic wrist and ankle, collar; Tyvek™ protective Fabric laminated with chemical-resistant Dow Saranex™ 23-P film | |
| 112.39 to 121.28 cm | |
| 38.25 in. | |
| 28.75 in. | |
| 1.82 to 1.95 m |