missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 4000 Coverall (all)

Catalog No. 18760C
Change view
Click to view available options
Seam Type:
Bound
Taped
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Seam Type Size
18760C Bound Large
19162778 Taped 5X-Large
19715335 Taped Large
189992583 Bound 4X-Large
19715334 Taped Medium
18760D Bound X-Large
18760E Bound 3X-Large
19162777 Taped 4X-Large
19715336 Taped 2X-Large
19087309 Bound 2X-Large
18760B Bound Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 18760C Supplier DuPont™ Supplier No. SL125BWHLG001200
Only null left

Useful when handling many wet and dry chemicals, and certain hazardous materials such as PCBs - See more at: http://beta.fishersci.com/shop/products//p-25013#sthash.pbR5bBoS.dpuf

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Laydown collar
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex

Compliance

Meets ASTM™ D3776, ASTM D1777, ASTM D3786, ASTM D5034 standards.

Specifications

Size Large
Gender Unisex
Material Tychem
Color White
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Closure Type Front Zipper
Seam Type Bound
Description Elastic wrist and ankle, collar; Tyvek™ protective Fabric laminated with chemical-resistant Dow Saranex™ 23-P film
Size (English) Inseam 28.5 in.
Size (Metric) Chest 89.53 to 98.42 cm
For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
Length (English) Sleeve 35 in.
Length (Metric) Sleeve 88.9 cm
Width (English) Chest 25.75 in.
Height (English) 5.3 to 5.7 ft.
Height (Metric) 1.6 to 1.73 m
Show More Show Less