missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 4000 Hooded Coverall

Catalog No. 29801436
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
29801436 Medium
29801437 Large
29801438 X-Large
29801439 2X-Large
19120694C 3X-Large
296356796 4X-Large
17500151 7X-Large
17500150 6X-Large
18999593 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 29801436 Supplier DuPont™ Supplier No. SL127BWHMD001200

DuPont™ Tychem™ 4000 Coveralls provide at least 30 minutes of protection against >124 chemical challenges in a range of chemical environments.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Attached hood with elastic around face opening.
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex

Compliance

Meets ASTM™ D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786; 16 CFR 1610 Standard for Flammability of Clothing Textiles; MIL-STD-282 requirements.

Specifications

Size Medium
Gender Unisex
Material Tychem
Color White
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Seam Type Bound
Certifications/Compliance ASTM D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786, 16 CFR 1610, MIL-STD-282
Size (English) Chest 35.25 to 38.75 in.
Size (English) Inseam 27.5 in.
Size (Metric) Chest 89.53 to 98.42 cm
Thickness 12 mil
For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
Length (English) Sleeve 33.75 in.
Length (Metric) Sleeve 85.72 cm
Width (English) Chest 24.25 in.
Height (English) 5.3 to 5.7 ft.
Height (Metric) 1.6 to 1.73 m
Weight (English) 0.22 lb.
Weight (Metric) 0.1 kg
Show More Show Less