missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 4000 Hooded Coverall

Catalog No. 19166136
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19166136 4X-Large
0136151B Medium
0136151C Large
0136151D X-Large
189992216 2X-Large
189992217 3X-Large
190881701 5X-Large
17500147 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19166136 Supplier DuPont™ Supplier No. SL122BWH4X001200

Tychem™ 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Attached hood with elastic around face opening.
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Manufactured under specifications that do not contain natural rubber latex

Compliance

Meets ASTM™ D3776, ASTM D1777, ASTM D3786, ASTM D5034 standards.

Specifications

Product Type Hooded Coveralls with Socks
Size 4X-Large
Gender Unisex
Material Tychem
Color White
Disposable Single-use
Ankle Style Attached Socks
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type Front Zipper
Seam Type Bound
Sterility Non-sterile
Description Attached hood, elastic wrist, attached socks
Size (English) Chest 50.75 to 54.25 in.
Size (English) Inseam 32 in.
Size (Metric) Inseam 81.28 cm
Thickness 12 mil
For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
Length (English) Sleeve 39.5 in.
Length (Metric) Sleeve 100.33 cm
Width (English) Chest 32 in.
Width (Metric) Chest 81.28 cm
Height (English) 6.33 to 6.58 ft.
Height (Metric) 1.92 to 2.01 m
Show More Show Less