missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 4000 Hooded Coverall
Tychem™ 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges.
Supplier: DuPont™ SL122BWH5X001200
Description
- Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
- Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
- Attached hood with elastic around face opening.
- Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Integrated socks composed of garment material
- Manufactured under specifications that do not contain natural rubber latex
Compliance
Meets ASTM™ D3776, ASTM D1777, ASTM D3786, ASTM D5034 standards.
Specifications
| Hooded Coveralls with Socks | |
| Unisex | |
| White | |
| Attached Socks | |
| Hooded | |
| Front Zipper | |
| Non-sterile | |
| 53 .75 to 57 .25 in. | |
| 83.82 cm | |
| Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments | |
| 102.87 cm | |
| 85.09 cm | |
| 2.01 to 2.07 m |
| 5X-Large | |
| Tychem | |
| Single-use | |
| Elastic | |
| Attached, Elastic Face | |
| Bound | |
| Attached hood, elastic wrist, attached socks | |
| 33 in. | |
| 12 mil | |
| 40.5 in. | |
| 33.5 in. | |
| 6.58 to 6.08 ft. |