Learn More
DuPont™ Tychem™ 4000 Coverall (all)
Catalog No. p-7139010
Change view
Click to view available options
Size:
3X-Large
4X-Large
5X-Large
6X-Large
Catalog No. Size
17500133 3X-Large
17500134 4X-Large
17500135 5X-Large
17500136 6X-Large
4 options
Catalog No. 17500133
Supplier: DuPont™ SL120BWH3X001200
Only null left
DuPont™ TYCHEM™ 4000 Coveralls provide at least 30 minutes of protection against >124 chemical challenges in a range of chemical environments.
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Laydown collar
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Open wrists
  • Open ankles
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Size 3X-Large
Gender Unisex
Material Tychem
Color White
Disposable Single-use
Ankle Style Open
Cuff Style Open
Neck Style Collared
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Resistance Chemicals, Bloodborne Pathogens
No. of Pockets None
Show More Show Less