Learn More
DuPont™ Tychem™ 2000 SFR Sleeved Apron
Catalog No. p-9045005
Change view
Click to view available options
Size:
2X-Large
Catalog No. Size
17986626 2X-Large
1 options
Catalog No. 17986626 Supplier DuPont™ Supplier No. QS275TGR2X001200
SFR Aprons provide chemical and secondary flame protection in a lightweight garment
  • Knee-length apron/gown design with attached sleeves.
  • Manufactured under specifications that do not contain natural rubber latex
  • Provides enhanced frontal protection.
  • 44 in. long for neck-to-knee coverage.
  • Open back for wearer comfort
  • Hook and loop closure.
  • Ties at waist.
  • Elastic opening for tighter fit at wrist.
  • Available in multiple sizes.
  • Taped seams provide strong chemical resistance against heavy liquid splashes
Specifications
Size 2X-Large
Material Tychem
Color Green
Disposable Single-use
Closure Type Waist Ties
Sterility Non-sterile
Cuff Style Elastic
Latex Free Yes
Thickness 7.5 mil
Length (English) 45.75 in.
Length (English) Cuff 33 in.
Length (Metric) 112 cm
Seam Type Taped
Show More Show Less