Learn More
DuPont™ Tychem™ 10000 Training Suit
Description
- Tychem™ 10000 fully encapsulated training suits offer an economical and effective option for HazMat training. Same fabric and garment design as standard Tychem™ 10000 suits
- Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
- Extra wide (EX) single layer faceshield (20 mil PVC) for wide viewing angle
- Front entry design allows wearer to partially don garment (maintain standby position) without assistance
- Integrated butyl gloves
- Internal adjustable waist belt system for support and improved fit
- Clearly labeled for training use only
- Manufactured under specifications that do not contain natural rubber latex
- Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
Specifications
Specifications
| Color | Lime Yellow |
| Material | Tychem |
| Size | 3X-Large |
| Ankle Style | Boot Overflaps |
| Closure Type | Zipper |
| Seam Type | Serged |