Learn More
DuPont™ Tychem™ 10000 Encapsulated Level A Suit with Viton™
Encapsulated Level A garment offers a high level of protection against toxic, corrosive gases and liquid chemicals
Supplier:  DuPont™ TK554TLY4X00015C
 View more versions of this product
Catalog No. 18888850
Only null left
Explore available promotions
Description
Description
  • 10000 Encapsulated Level A Suit with Viton™ design provides protection from liquid splash exposure for both the wearer and respiratory equipment
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Manufactured under specifications that do not contain natural rubber latex
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • Also provides coverage from overhead liquid splash (does not provide vapor protection)
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Standard 40 mil PVC faceshield
  • Front entry design allows wearer to partially don garment (maintain standby position) without assistance
  • Expanded back to accommodate self-contained air breathing apparatus (SCBA)
  • Liquid resistant, extra long zipper to increase garment opening and aid donning and doffing
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
Specifications
Specifications
Lime Yellow
4X-Large
Zipper
Tychem
Boot Overflaps
Taped
Product Suggestions
Product Suggestions