Learn More
DuPont™ Tychem™ 10000 Encapsulated Level A Suit with MSA Dual Purpose
Catalog No. p-7139609
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18888859 2X-Large
18888869 Large
18888876 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18888859 Supplier DuPont™ Supplier No. TK555TLY2X00017M
Only null left
Suit with MSA dual purpose offers a high level of protection against toxic, corrosive gases and liquid chemicals
  • 10000 Encapsulated Level A Suit with MSA Dual Purpose garment design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Two exhaust valves release higher pressure inside encapsulating garments while simultaneously helping to prevent inward leakage of external vapors or particles into the garment
  • Internal adjustable waist belt system for support and improved fit
  • Additional barrier material sewn at knees and over taped for added abrasion resistance when kneeling or crawling
  • Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052
  • Manufactured under specifications that do not contain natural rubber latex
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • Rear entry design keeps closure away from direct contact with frontal chemical exposure and provides ability to change air bottle on SCBA without removing the entire garment
  • Expanded back to accommodate self-contained air breathing apparatus (SCBA)
  • Vapor tight zipper
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Attached internal gloves composed of five layer laminate film for a broad range of chemical protection
  • Butyl gloves provide mechanical and physical durability to help protect inner glove
  • Gloves are attached with vapor tight ring and clamp system and are replaceable
Specifications
Color Lime Yellow
Material Tychem
Size 2X-Large
Ankle Style Boot Overflaps
Closure Type Zipper
Seam Type Taped