Learn More
DuPont™ Tychem™ 10000 Encapsulated Level A Suit with MSA Dual Purpose
Suit with MSA dual purpose offers a high level of protection against toxic, corrosive gases and liquid chemicals
Supplier:  DuPont™ TK555TLYLG00017M
 View more versions of this product
Catalog No. 18-888-869
Only null left
Explore available promotions
Description
Description
  • 10000 Encapsulated Level A Suit with MSA Dual Purpose garment design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Two exhaust valves release higher pressure inside encapsulating garments while simultaneously helping to prevent inward leakage of external vapors or particles into the garment
  • Internal adjustable waist belt system for support and improved fit
  • Additional barrier material sewn at knees and over taped for added abrasion resistance when kneeling or crawling
  • Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052
  • Manufactured under specifications that do not contain natural rubber latex
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • Rear entry design keeps closure away from direct contact with frontal chemical exposure and provides ability to change air bottle on SCBA without removing the entire garment
  • Expanded back to accommodate self-contained air breathing apparatus (SCBA)
  • Vapor tight zipper
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Attached internal gloves composed of five layer laminate film for a broad range of chemical protection
  • Butyl gloves provide mechanical and physical durability to help protect inner glove
  • Gloves are attached with vapor tight ring and clamp system and are replaceable
Specifications
Specifications
Lime Yellow
Large
Zipper
Tychem
Boot Overflaps
Taped
Product Suggestions
Product Suggestions