Learn More
DuPont™ Tychem™ 10000 Encapsulated Level A Suit with Hansen Fitting
Encapsulated Level A Suit with hansen fitting offers a high level of protection against toxic, corrosive gases and liquid chemicals
Supplier:  DuPont™ TK554TLY3X00017S
 View more versions of this product
Catalog No. 18888849
Only null left
Explore available promotions
Description
Description
  • Encapsulated Level A Suit with hansen fitting design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052
  • Manufactured under specifications that do not contain natural rubber latex
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • Double taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered, both inside and outside of garment with compatible chemical-resistant material by heat-sealing.
  • Expanded anti-fog faceshield provides wide 220 degree viewing angle. EX faceshield is three-layer laminate composed of 40 mil PVC/5 mil Teflon™ / 20 mil PVC
  • Front entry design allows wearer to partially don garment (maintain standby position) without assistance
  • Expanded back to accommodate self-contained air breathing apparatus (SCBA)
  • Vapor tight zipper. Extra long to increase garment opening and aid donning and doffing
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Attached internal gloves composed of five layer laminate film for a broad range of chemical protection
  • Butyl gloves provide mechanical and physical durability to help protect inner glove Gloves are attached with vapor tight ring and clamp system and are replaceable
Specifications
Specifications
Lime Yellow
3X-Large
Zipper
Tychem
Boot Overflaps
Taped
Product Suggestions
Product Suggestions