Learn More
DuPont™ Tychem™ 10000 Encapsulated Level A Suit with Hansen Fitting
Catalog No. 18888847
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18888847 2X-Large
18888849 3X-Large
18888851 4X-Large
18888853 Large
18888855 Medium
18888856 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18888847 Supplier DuPont™ Supplier No. TK554TLY2X00017S
Encapsulated Level A Suit with hansen fitting offers a high level of protection against toxic, corrosive gases and liquid chemicals
  • Encapsulated Level A Suit with hansen fitting design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052
  • Manufactured under specifications that do not contain natural rubber latex
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • Double taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered, both inside and outside of garment with compatible chemical-resistant material by heat-sealing.
  • Expanded anti-fog faceshield provides wide 220 degree viewing angle. EX faceshield is three-layer laminate composed of 40 mil PVC/5 mil Teflon™ / 20 mil PVC
  • Front entry design allows wearer to partially don garment (maintain standby position) without assistance
  • Expanded back to accommodate self-contained air breathing apparatus (SCBA)
  • Vapor tight zipper. Extra long to increase garment opening and aid donning and doffing
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Attached internal gloves composed of five layer laminate film for a broad range of chemical protection
  • Butyl gloves provide mechanical and physical durability to help protect inner glove Gloves are attached with vapor tight ring and clamp system and are replaceable
Specifications
Color Lime Yellow
Material Tychem
Size 2X-Large
Ankle Style Boot Overflaps
Closure Type Zipper
Seam Type Taped